missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOLA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15701432 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15701432 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15701432 Supplier Invitrogen™ Supplier No. PA563656

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GRGGFNKGQD QGPPERVVLL GEFLHPCEDD IVCKCTT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Required for ribosome biogenesis and telomere maintenance. NOLA1 is part of the H/ACA small nucleolar ribonucleoprotein (H/ACA snoRNP) complex, which catalyzes pseudouridylation of rRNA. This involves the isomerization of uridine such that the ribose is subsequently attached to C5, instead of the normal N1. Each rRNA can contain up to 100 pseudouridine ('psi') residues, which may serve to stabilize the conformation of rRNAs. NOLA1 may also be required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOLA1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAR1
Gene Accession No. Q9NY12
Gene Alias AA409823; AI326794; C430047J18Rik; Gar1; GAR1 homolog, ribonucleoprotein; GAR1 ribonucleoprotein; GAR1 ribonucleoprotein homolog; GAR1 ribonucleoprotein homolog (yeast); H/ACA ribonucleoprotein complex subunit 1; H/ACA small nucleolar RNPs; Nola1; Nucleolar protein family A member 1; nucleolar protein family A, member 1 (H/ACA small nucleolar RNPs); snoRNP protein GAR1
Gene Symbols GAR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOLA1. Recombinant protein control fragment (Product #RP-101734).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.