missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575894
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Proteins that contain MIF4G (middle of eIF4G (MIM 600495)) and/or MA3 domains, such as NOM1, function in protein translation. These domains include binding sites for members of the EIF4A family of ATP-dependent DEAD box RNA helicases (see EIF4A1; MIM 602641) (Simmons et al., 2005 [PubMed 15715967]).[supplied by OMIM, Mar 2008].
Specifications
| NOM1 | |
| Polyclonal | |
| Unconjugated | |
| NOM1 | |
| C7orf3; D5Kng1; Gm1040; Nom1; Nucleolar MIF4G domain-containing protein 1; nucleolar protein with MIF4G domain 1; PPP1R113; protein phosphatase 1, regulatory subunit 113; SGD1; SGD1 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64434, 682058 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5C9Z4 | |
| NOM1 | |
| Synthetic peptide corresponding to residues in Human NOM1. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction