missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539315
This item is not returnable.
View return policy
Description
Proteins that contain MIF4G (middle of eIF4G (MIM 600495)) and/or MA3 domains, such as NOM1, function in protein translation. These domains include binding sites for members of the EIF4A family of ATP-dependent DEAD box RNA helicases (see EIF4A1; MIM 602641) (Simmons et al., 2005 [PubMed 15715967]).[supplied by OMIM, Mar 2008].
Specifications
| NOM1 | |
| Polyclonal | |
| Unconjugated | |
| NOM1 | |
| Gm1040, C7orf3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 433864, 64434 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5C9Z4, Q3UFM5 | |
| NOM1 | |
| A synthetic peptide derived from the C-terminal region of human NOM1 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction