missing translation for 'onlineSavingsMsg'
Learn More

Nomifensine maleate, MedChemExpress

Product Code. 16454547
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16454547

Brand: Medchem Express HYB1110A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nomifensine maleate is a selective inhibitor of dopamine uptake, used in adult attention deficit disorder.

Nomifensine maleate is a selective inhibitor of dopamine uptake, used in adult attention deficit disorder.

Specifications

Chemical Name or Material Nomifensine maleate
CAS 32795-47-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H22N2O4
Quantity 50 mg
Synonym (±)-Nomifensine maleat
Solubility Information DMSO : ≥ 80 mg/mL (225.73 mM) ∣H2O : 2.2 mg/mL (6.21 mM; Need ultrasonic)
SMILES NC1=CC=CC2=C1CN(C)CC2C3=CC=CC=C3.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 354.4
Formula Weight 354.4
Percent Purity 99.67%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.