Learn More
Nonadecane, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | n-Nonadecane |
| CAS | 629-92-5 |
| Molecular Formula | C19 H40 |
| Quantity | 10 g |
| Synonym | Nonadecane, NSC 77136, n-Nonadecane |
| InChI Formula | InChI=1S/C19H40/c1-3-5-7-9-11-13-15-17-19-18-16-14-12-10-8-6-4-2/h3-19H2,1-2H3 |
| SMILES | CCCCCCCCCCCCCCCCCCC |
| IUPAC Name | nonadecane |
| Molecular Weight (g/mol) | 268.52 |
| Formula Weight | 268.313 |