missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NONO Antibody (OTI3F8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73033FR
This item is not returnable.
View return policy
Description
NONO Monoclonal specifically detects NONO in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| NONO | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| NONO | |
| Recombinant protein fragment corresponding to amino acids 184-385 of human NONO (NP_031389) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3F8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| NMT5552 kDa subunit, non-POU domain containing, octamer-binding, NRB54non-POU-domain-containing, octamer-binding, p54(nrb) | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 4841.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction