missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NONO Antibody (OTI4D9), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73034MFV610
This item is not returnable.
View return policy
Description
NONO Monoclonal antibody specifically detects NONO in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry
Specifications
| NONO | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Flow Cytometry | |
| OTI4D9 | |
| Flow Cytometry | |
| NMT5552 kDa subunit, non-POU domain containing, octamer-binding, NRB54non-POU-domain-containing, octamer-binding, p54(nrb) | |
| Human recombinant protein fragment correspongding to amino acids 184-385 of human NONO(NP_031389) produced in E.coli. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 4841 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction