Learn More
Invitrogen™ NOP1 Monoclonal Antibody (28F2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110025
Description
MA1-10025 detects Yeast NOP-1 from S. cerevisiae samples and does not cross-react with fibrillarin in other species. For chemiluminescent Western blot using yeast samples, use a dilution of 1:10,000 for cell lysates and 1:25,000 for nuclear fractions. For non-ECL detection methods, use a dilution of 1:1000-1:5000.
Nop1p is the yeast homologue of human fibrillarin, a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle.
Specifications
| NOP1 | |
| Monoclonal | |
| Conc. Not Determined | |
| Tissue culture supernatant with 5mM sodium azide | |
| P15646 | |
| NOP1 | |
| Yeast nuclear preparation. | |
| RUO | |
| 851548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 28F2 | |
| Unconjugated | |
| NOP1 | |
| D2870; Histone-glutamine methyltransferase; LOT3; NOP1; Nop1p; Nucleolar protein 1; rRNA 2'-O-methyltransferase fibrillarin; rRNA methyltransferase NOP1; U3 small nucleolar RNA-associated protein NOP1; YDL014W | |
| Mouse | |
| 500 μL | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.