missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP14 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22227AF532
This item is not returnable.
View return policy
Description
NOP14 Polyclonal antibody specifically detects NOP14 in Human, Mouse samples. It is validated for Western Blot
Specifications
| NOP14 | |
| Polyclonal | |
| Western Blot | |
| C4orf9, chromosome 4 open reading frame 9, NOL14nucleolar protein 14 homolog (yeast), NOP14 homolog, NOP14 nucleolar protein homolog (yeast), Nucleolar complex protein 14, nucleolar protein 14, nucleolar protein 14 homolog, probable nucleolar complex protein 14, RES425, RES4-25 | |
| The immunogen this antibody was made to, maps to a region between residue 750 and 800 of human Nucleolar Complex Protein 14 using the numbering given in entry NP_003694.1 (GeneID 8602). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8602 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction