missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOP14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103985
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NOL14.
Involved in nucleolar processing of pre-18S ribosomal RNA. Has a role in the nuclear export of 40S pre-ribosomal subunit to the cytoplasm.
Specifications
| NOP14 | |
| Polyclonal | |
| Unconjugated | |
| NOP14 | |
| 2610033H07Rik; C4orf9; NOL14; NOP14; NOP14 nucleolar protein; NOP14 nucleolar protein homolog; nucleolar complex protein 14; Nucleolar protein 14; probable nucleolar complex protein 14; RES425; RES4-25; RGD1305605; UTP2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 289724, 75416, 8602 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P78316, Q8R3N1 | |
| NOP14 | |
| A synthesized peptide derived from human NOP14(Accession P78316), corresponding to amino acid residues M338-S388. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction