missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nope/IGDCC4 Antibody (2516A) [DyLight 755], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10345Z
This item is not returnable.
View return policy
Description
Nope/IGDCC4 Monoclonal antibody specifically detects Nope/IGDCC4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Nope/IGDCC4 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 57722 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 2516A | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| DDM36, hDDM36, immunoglobulin superfamily DCC subclass member 4, immunoglobulin superfamily, DCC subclass, member 4, KIAA1628, likely ortholog of mouse neighbor of Punc E11, LOC57722, Neighbor of punc e11, NOPEFLJ42051, Protein DDM36 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Nope/IGDCC4, Met1-Thr957, Accession # Q8TDY8 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction