missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NORE1 Antibody (OTI1H2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03691
This item is not returnable.
View return policy
Description
NORE1 Monoclonal specifically detects NORE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| NORE1 | |
| Monoclonal | |
| 0.41 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:100 | |
| RASSF5 | |
| Full length human recombinant protein of human RASSF5 (NP_872606) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| OTI1H2 | |
| Unconjugated | |
| Maxp1, MGC10823, MGC17344, New ras effector 1, NORE1A, NORE1B, NORE1RASSF3, Rap1-binding protein, RAPLnovel Ras effector 1, Ras association (RalGDS/AF-6) domain family 5, Ras association (RalGDS/AF-6) domain family member 5, ras association domain-containing protein 5, Ras effector-like protein, regulator for cell adhesion and polarization enriched in lymphoid tissue, Regulator for cell adhesion and polarization enriched in lymphoid tissues, tumor suppressor RASSF3 | |
| Mouse | |
| 30.2 kDa | |
| 0.1 mL | |
| Cancer, Signal Transduction, Tumor Suppressors | |
| 83593 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction