missing translation for 'onlineSavingsMsg'
Learn More

Norethisterone Acetate, Mikromol™

Product Code. 18080970
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 18080970

Brand: Mikromol MM0219.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Norethisterone Acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norethisterone acetate
CAS 51-98-9
Analyte or Component Names Norethisterone acetate
Shipping Condition Room Temperature
Molecular Formula C22 H28 O3
Certifications/Compliance ISO 17034
Synonym Norethisterone Acetate, 19-Norethisterone acetate, 19-Norethynyltestosterone acetate, Aygestin, Gynophase, Gynovlane, Milli-Anovlar, Milligynon, Miniphase, NSC 22844, Norethindrone 17-acetate, Norethindrone acetate, Norethisteron acetate, Norethisterone 17-acetate, Norethisterone acetate, Norethynyltestosterone acetate, Norethysterone acetate, Norlutate, Norlutin A, Norlutin acetate, Orlutate, Primolut-Nor, Progylut, SH 420, (17alpha)-17-(Acetyloxy)-19-norpregn-4-en-20-yn-3-one, 17-Hydroxy-19-nor-17alpha-pregn-4-en-20-yn-3-one acetate, 17-Acetoxy-19-nor-17alpha-pregn-4-en-20-yn-3-one, 17alpha-Ethinyl-19-nortestosterone 17beta-acetate, 17alpha-Ethinyl-19-nortestosterone acetate, 17alpha-Ethinyl-4-estrene-17beta-ol-acetate-3-one, 17alpha-Ethynyl-19-nortestosterone acetate, 17beta-Acetoxy-17alpha-ethinyl-4-estren-3-one, 17beta-Acetoxy-19-nor-17alpha-pregn-4-en-20-yn-3-one, 17beta-Hydroxy-19-nor-17alpha-pregn-4-en-20-yn-3-one acetate
InChI Formula InChI=1S/C22H28O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,13,17-20H,5-12H2,2-3H3/t17-,18+,19+,20-,21-,22-/m0/s1
SMILES CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 340.46
Quantity 250 mg
Formula Weight 340.2038 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.