Learn More
Norethisterone Acetate (Norethindrone Acetate), LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Norethisterone acetate |
| CAS | 51-98-9 |
| Analyte or Component Names | Norethisterone acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Norethisterone Acetate, 19-Norethisterone acetate, 19-Norethynyltestosterone acetate, Aygestin, Gynophase, Gynovlane, Milli-Anovlar, Milligynon, Miniphase, NSC 22844, Norethindrone 17-acetate, Norethindrone acetate, Norethisteron acetate, Norethisterone 17-acetate, Norethisterone acetate, Norethynyltestosterone acetate, Norethysterone acetate, Norlutate, Norlutin A, Norlutin acetate, Orlutate, Primolut-Nor, Progylut, SH 420, (17alpha)-17-(Acetyloxy)-19-norpregn-4-en-20-yn-3-one, 17-Hydroxy-19-nor-17alpha-pregn-4-en-20-yn-3-one acetate, 17-Acetoxy-19-nor-17alpha-pregn-4-en-20-yn-3-one, 17alpha-Ethinyl-19-nortestosterone 17beta-acetate, 17alpha-Ethinyl-19-nortestosterone acetate, 17alpha-Ethinyl-4-estrene-17beta-ol-acetate-3-one, 17alpha-Ethynyl-19-nortestosterone acetate, 17beta-Acetoxy-17alpha-ethinyl-4-estren-3-one, 17beta-Acetoxy-19-nor-17alpha-pregn-4-en-20-yn-3-one, 17beta-Hydroxy-19-nor-17alpha-pregn-4-en-20-yn-3-one acetate |
| InChI Formula | InChI=1S/C22H28O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,13,17-20H,5-12H2,2-3H3/t17-,18+,19+,20-,21-,22-/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C |
| Show More |