missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Normal Sheep IgG, APC Negative Control, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems IC016A
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Control.
Specifications
| IgG | |
| APC | |
| Immunoglobulin G, ImmunoglobulinG | |
| Sheep | |
| Protein A or G purified | |
| 3500 | |
| Sheep | |
| IgG |
| Polyclonal | |
| Control 10 uL/10^6 cells | |
| IGHG1 | |
| NA | |
| 200 Tests | |
| Serum was obtained from naive (non-immunized) sheep. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu