missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Norovirus capsid G2 Monoclonal Antibody (F29H)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518242
This item is not returnable.
View return policy
Description
This antibody does not cross-react with capsid G1. Purity is >90%. Recommended pairs for ELISA: MA5-18241 (capture) and MA5-18242 (conjugated, detection).
Specifications
| Norovirus capsid G2 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA | |
| F29H | |
| Unconjugated | |
| Norwalk virus | |
| Norovirus recombinant protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction