missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norrin/NDP Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77014AF594
This item is not returnable.
View return policy
Description
Norrin/NDP Polyclonal antibody specifically detects Norrin/NDP in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Norrin/NDP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 594 | |
| EVR2, exudative vitreoretinopathy 2 (X-linked), FEVR, ND, Norrie disease (pseudoglioma), Norrie disease protein, norrin, X-linked exudative vitreoretinopathy 2 protein | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the amino terminus of human Norrin. The immunogen is located within the first 50 amino acids of Norrin. Amino Acid Squence: DTDSKTDSSFIMDSDPRR | |
| 0.1 mL | |
| Cell Cycle and Replication, Neuroscience, Vision | |
| 4693 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction