missing translation for 'onlineSavingsMsg'
Learn More

Nortriptyline Hydrochloride, TRC

Product Code. 30135249
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135249 1 g 1g
30138652 100 mg 100mg
17930685 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135249 Supplier Toronto Research Chemicals Supplier No. TRCN8370001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Formula Weight 299.1441
Molecular Formula C19 H21 N . Cl H
Molecular Weight (g/mol) 299.84
Chemical Name or Material Nortriptyline hydrochloride

Specifications

CAS 894-71-3
Shipping Condition Room Temperature
Synonym Nortriptyline Hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)-N-methylpropan-1-amine Hydrochloride, Amitriptyline Hydrochloride Imp. C (EP) as Hydrochloride, 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-, hydrochloride (1:1), 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-, hydrochloride (9CI), 5H-Dibenzo[a,d]cycloheptene-δ5,γ-propylamine, 10,11-dihydro-N-methyl-, hydrochloride (8CI), 10,11-Dihydro-N-methyl-5H-dibenzo[a,d]cycloheptene-δ5,γ-propylamine hydrochloride, 38489, 5-(3-Methylaminopropylidene)-10,11-dihydro-5H-dibenzo[a,d]cycloheptene hydrochloride, 5-[3-(Methylamino)propylidene]dibenzo[a,e]cyclohepta[1,5]diene hydrochloride, Acetexa, Allegron, Altilev, Aventyl hydrochloride, Desmethylamitriptyline hydrochloride, Noramitriptyline hydrochloride, Norfenazin, Noritren, Nortrilen, Nortryptilinium hydrochloride, Norzepine, Pamelor, Psychostyl, Sensival, Vividyl
InChI Formula InChI=1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-11,20H,6,12-14H2,1H3;1H
SMILES Cl.CNCCC=C1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N-methylpropan-1-amine;hydrochloride
Industry Type Pharmaceutical
Quantity 1 g
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.