missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOS1AP Monoclonal Antibody (OTI10F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811699
This item is not returnable.
View return policy
Description
This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| NOS1AP | |
| Monoclonal | |
| Unconjugated | |
| NOS1AP | |
| 6330408P19Rik, CAPON | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9722 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI10F8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75052 | |
| NOS1AP | |
| Human recombit protein fragment corresponding to amino acids 1-196 of human NOS1AP produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction