missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOS1AP Monoclonal Antibody (OTI10F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811699S
This item is not returnable.
View return policy
Description
NOS1AP Monoclonal antibody specifically detects NOS1AP in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| NOS1AP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8BHJ5, Q9BZK7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 365755, 79718, 81004 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI10F8 | |
| Unconjugated | |
| TBL1XR1 | |
| 6330408P19Rik; Capon; carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein; C-terminal PDZ domain ligand of neuronal nitric oxide synthase; C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON); C-terminal PDZ ligand of neuronal nitric oxide synthase protein; Kiaa0464; ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain; MGC138500; nitric oxide synthase 1 (neuronal) adaptor protein; nitric oxide synthase 1 adaptor protein; NOS1AP | |
| Full length human recombinant protein of human TBL1XR1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction