missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOSIP Polyclonal Antibody
GREENER_CHOICE

Product Code. 13259458
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
13259458 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 13259458 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PA522206

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NEDD4L is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. NEDD4L inhibits TGF-beta signaling by triggering SMAD2 and TGFR1 ubiquitination and proteasome-dependent degradation. NEDD4L promotes ubiquitination and internalization of various plasma membrane channels such as ENaC, Nav1. 2, Nav1. 3, Nav1. 5, Nav1. 7, Nav1. 8, Kv1. 3, EAAT1 or CLC5. NEDD4L also promotes ubiquitination and degradation of SGK.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen NOSIP
Applications Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene NOSIP
Gene Accession No. Q9Y314
Gene Alias 2310061K06Rik; CGI-25; E3 ubiquitin-protein ligase NOSIP; eNOS-interacting protein; nitric oxide synthase interacting protein; nitric oxide synthase-interacting protein; Nosip; RING-type E3 ubiquitin transferase NOSIP
Gene Symbols NOSIP
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human NOSIP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51070
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.