missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-1 Antibody (E6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 282.00 - € 602.00
Specifications
| Antigen | Notch-1 |
|---|---|
| Clone | E6 |
| Dilution | Flow Cytometry, ELISA 1:100 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18054534
|
Novus Biologicals
NBP2-62557-0.025MG |
0.025 mg |
€ 282.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610648
|
Novus Biologicals
NBP2-62557 |
0.2 mg |
€ 602.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Notch-1 Monoclonal specifically detects Notch-1 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence.Specifications
| Notch-1 | |
| Flow Cytometry, ELISA 1:100 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| PBS | |
| 4851 | |
| NRR of mouse Notch1. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| E6 | |
| Monoclonal | |
| Purified | |
| Human, Mouse | |
| EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
| NOTCH1 | |
| IgG λ | |
| An engineered ScFv fragment antibody that binds to both mouse mNRR1 and human hNRR1 but not NRR2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title