missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-1 Antibody (N1A), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5267B
This item is not returnable.
View return policy
Description
Notch-1 Monoclonal antibody specifically detects Notch-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Notch-1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| N1A | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
| Mouse embryonic fibroblast cell line PA317-derived recombinant mouse Notch-1, Accession # Q01705 | |
| 0.1 mL | |
| Apoptosis, Cancer Stem Cells, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Signaling Pathway | |
| 4851 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction