missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-4 Antibody (S08-8H5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19822-50ul
This item is not returnable.
View return policy
Description
Notch-4 Monoclonal antibody specifically detects Notch-4 in Human samples. It is validated for Western Blot
Specifications
| Notch-4 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S08-8H5 | |
| Unconjugated | |
| neurogenic locus notch homolog protein 4, notch 4, Notch homolog 4, NOTCH3 | |
| A synthetic peptide of human Notch-4 (Uniprot # Q99466) | |
| 50 μg | |
| Angiogenesis | |
| 4855 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction