missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Noto Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549419
This item is not returnable.
View return policy
Description
Noto Polyclonal Antibody for Western Blot
Transcription regulator acting downstream of both FOXA2 and T during notochord development. Required for node morphogenesis. Is essential for cilia formation in the posterior notochord (PNC) and for left-right patterning; acts upstream of FOXJ1 and RFX3 in this process and is required for the expression of various components important for axonemal assembly and function. Plays a role in regulating axial versus paraxial cell fate.
Specifications
| Noto | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5TIS6 | |
| Noto | |
| KLH conjugated synthetic peptide between 200-234 amino acids from the C-terminal region of Mouse Noto. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Noto | |
| Flh; floating head homolog; homeobox protein notochord; MmNot; Not; Not homeodomain protein; NOTO; notochord homeobox; notochord homolog; notochord homolog (Xenopus laevis); tc; truncate | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 384452 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction