missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AG-2/AGR2 Antibody Blocking Peptide

Product Code. 18231210 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18231210

Brand: Novus Biologicals™ NBP105936PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
A peptide to AGR2.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 10551
Species Human
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation Peptide dissolved in dH2O. Contains no BSA.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-05936. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog
Gene Symbol AGR2
Product Type AG-2
Quantity 0.05 mg
Regulatory Status RUO - research use only
Target Enzyme AG-2/AGR2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.