Learn More
Novus Biologicals™ alpha Satellite Repeat Primer
Product Code. 18202451 Shop All Bio Techne Products
Click to view available options
Quantity:
0.2 mL
0.5 mL
Unit Size:
0.2mL
0.5mL
Product Code. 18202451
Brand: Novus Biologicals™ NBP171654SS
alpha SAT primer for PCR in chromatin precipitation
  • Primer sets come premixed with forward and reverse primers at a concentration of 10μM
  • Primers are compatible with all real-time PCR chemistries
  • Each primer set has been tested and validated by chromatin immunoprecipitation (ChIP)
  • Tm for primer set is between 53.4° to 54°c
  • Amplicon size of primer set is 150 base pairs
Recommended Storage
Store at -20°C. Avoid freeze-thaw cycles.
TRUSTED_SUSTAINABILITY
Specifications
Product Type alpha Satellite Repeat Primer
Content And Storage Store at –20°C. Avoid Free/Thaw Cycles
Quantity 0.2 mL
For Use With (Application) Chromatin Immunoprecipitation