missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ATG16L1 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
A peptide to ATG16L1.
Specifications
Specifications
| Gene ID (Entrez) | 55054 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-60928. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30 |
| Gene Symbol | ATG16L1 |
| Product Type | ATG16L1 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction