missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HeLa CoCl2 treated/Untreated Cell Lysate

Product Code. 18775053 Shop All Bio Techne Products
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 18775053

Brand: Novus Biologicals™ NBP236450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The HeLa Hypoxic (CoCl2)/ Normoxic Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Cervix
Quantity 2 Vials
Storage Buffer The protein lysate is prepared in 1X Laemmli sample buffer with BME. Boiling before loading is not necessary.
Cellular Fraction Hypoxic (CoCl2) / Normoxic
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.