missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human DEFB116 His Protein

Product Code. 18198471 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18198471

Brand: Novus Biologicals™ NBP222992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 24-102 of Human DEFB116 The Recombinant Human DEFB116 Protein is derived from E. coli. The Recombinant Human DEFB116 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001032820
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 245930
Molecular Weight (g/mol) 11.5kDa
Name DEFB116 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen DEFB116, 24-102aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSGLFRSHN GKSREPWNPC ELYQGMCRNA CREYEIQYLT CPNDQKCCLK LSVKITSSKN VKEDYDSNSN LSVTNSSSYS HI
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Immunology
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.