missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human DERP6 His Protein

Product Code. 18147961 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18147961

Brand: Novus Biologicals™ NBP223019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-316 of Human DERP6 The Recombinant Human DERP6 Protein is derived from E. coli. The Recombinant Human DERP6 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_056177
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 23587
Molecular Weight (g/mol) 37.2kDa
Name DERP6 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen ELP5, 1-316aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMTPSEGA RAGTGRELEM LDSLLALGGL VLLRDSVEWE GRSLLKALVK KSALCGEQVH ILGCEVSEEE FREGFDSDIN NRLVYHDFFR DPLNWSKTEE AFPGGPLGAL RAMCKRTDPV PVTIALDSLS WLLLRLPCTT LCQVLHAVSH QDSCPGDSSS VGKVSVLGLL HEELHGPGPV GALSSLAQTE VTLGGTMGQA SAHILCRRPR QRPTDQTQWF SILPDFSLDL QEGPSVESQP YSDPHIPPVD PTTHLTFNLH LSKKEREARD SLILPFQFSS EKQQALLRPR PGQATSHIFY EPDAYDDLDQ EDPDDDLDI
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.