missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Dysbindin His Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-270 of Human Dysbindin The Recombinant Human Dysbindin Protein is derived from E. coli. The Recombinant Human Dysbindin Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_898862 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 0.5mM DTT, 20% glycerol |
| Gene ID (Entrez) | 84062 |
| Molecular Weight (g/mol) | 34.6kDa |
| Name | Dysbindin Protein |
| Purification Method | Affinity purified |
| Immunogen | MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMLS AHWEKKKTSL VELQEQLQQL PALIADLESM TANLTHLEAS FEEVENNLLH LEDLCGQCEL ERCKHMQSQQ LENYKKNKRK ELETFKAELD AEHAQKVLEM EHTQQMKLKE RQKFFEEAFQ QDMEQYLSTG YLQIAERREP IGSMSSMEVN VDMLEQMDLM DISDQEALDV FLNSGGEENT VLSPALGPES STCQNEITLQ VPNPSELRAK PPSSSSTCTD SATRDISEGG ESPVVQSDEE EVQVDTALAT SHTDREATPD GGEDSDS |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction