Learn More
Novus Biologicals™ Recombinant Human GRP78/HSPA5 His Protein
Product Code. 18785463 Shop All Bio Techne Products
Product Code. 18785463
Brand: Novus Biologicals™ NBC118378
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 20-650 of Human GRP78/HSPA5 The Recombinant Human GRP78/HSPA5 Protein is derived from E. coli. The Recombinant Human GRP78/HSPA5 Protein has been validated for the following applications: SDS-Page.