missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human IGFBP-rp1/IGFBP-7 His Protein

Product Code. 18167981 Shop All Bio Techne Products
Click to view available options
Quantity:
0.02 mg
Unit Size:
0.02mg
This item is not returnable. View return policy

Product Code. 18167981

Brand: Novus Biologicals™ NBP223142

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 27-282 of Human IGFBP-rp1/IGFBP-7 The Recombinant Human IGFBP-rp1/IGFBP-7 Protein is derived from E. coli. The Recombinant Human IGFBP-rp1/IGFBP-7 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001544
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 50% glycerol, 2mM DTT, 1mM EDTA
Gene ID (Entrez) 3490
Molecular Weight (g/mol) 28.8kDa
Name IGFBP-rp1/IGFBP-7 Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen IGFBP7, 27-282aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSSSSDTCG PCEPASCPPL PPLGCLLGET RDACGCCPMC ARGEGEPCGG GGAGRGYCAP GMECVKSRKR RKGKAGAAAG GPGVSGVCVC KSRYPVCGSD GTTYPSGCQL RAASQRAESR GEKAITQVSK GTCEQGPSIV TPPKDIWNVT GAQVYLSCEV IGIPTPVLIW NKVKRGHYGV QRTELLPGDR DNLAIQTRGG PEKHEVTGWV LVSPLSKEDA GEYECHASNS QGQASASAKI TVVDALHEIP VKKGEGAEL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.