missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human ISG15/UCRP Calmodulin Protein

Product Code. 18211471 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18211471

Brand: Novus Biologicals™ NBC118442

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-Terminal CaM-tag and corresponding to the amino acids 1-157 of Human ISG15/UCRP The Recombinant Human ISG15/UCRP Protein is derived from E. coli. The Recombinant Human ISG15/UCRP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05161
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris buffer (pH 8.0), 0.1 M NaCl, 1mM DTT
Gene ID (Entrez) 9636
Molecular Weight (g/mol) 34kDa
Name ISG15/UCRP Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen ISG15, 1-157 aa, Human, Calmodulin tagged, Recombinant, E.coli (NP_005092).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.