missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human LIN-28B His Protein

Product Code. 18116265 Shop All Bio Techne Products
Click to view available options
Quantity:
0.01 mg
Unit Size:
0.01mg
This item is not returnable. View return policy

Product Code. 18116265

Brand: Novus Biologicals™ NBP223196

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-250 of Human LIN-28B The Recombinant Human LIN-28B Protein is derived from E. coli. The Recombinant Human LIN-28B Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001004317
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 10% glycerol, 2mM DTT
Gene ID (Entrez) 389421
Molecular Weight (g/mol) 29.5kDa
Name LIN-28B Protein
Purification Method SDS-PAGE
Quantity 0.01 mg
Source E. Coli
Immunogen Lin28B, 1-250aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAEGGAS KGGGEEPGKL PEPAEEESQV LRGTGHCKWF NVRMGFGFIS MINREGSPLD IPVDVFVHQS KLFMEGFRSL KEGEPVEFTF KKSSKGLESI RVTGPGGSPC LGSERRPKGK TLQKRKPKGD RCYNCGGLDH HAKECSLPPQ PKKCHYCQSI MHMVANCPHK NVAQPPASSQ GRQEAESQPC TSTLPREVGG GHGCTSPPFP QEARAEISER SGRSPQEASS TKSSIAPEEQ SKKGPSVQKR KKT
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.