missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human MafF His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-164 of Human MafF The Recombinant Human MafF Protein is derived from E. coli. The Recombinant Human MafF Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_001155045 |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.4M UREA |
| Gene ID (Entrez) | 23764 |
| Molecular Weight (g/mol) | 20.1kDa |
| Name | MafF Protein |
| Purification Method | SDS-PAGE |
| Quantity | 0.1 mg |
| Source | E. Coli |
| Immunogen | MAFF, 1-164aa, Human His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSVDPLS SKALKIKREL SENTPHLSDE ALMGLSVREL NRHLRGLSAE EVTRLKQRRR TLKNRGYAAS CRVKRVCQKE ELQKQKSELE REVDKLAREN AAMRLELDAL RGKCEALQGF ARSVAAARGP ATLVAPASVI TIVKSTPGSG SGPAHGPDPA HGPASCS |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction