missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CD43/Sialophorin His Protein

Product Code. 18129991 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18129991

Brand: Novus Biologicals™ NBP223472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-253 of Human Sialophorin The Recombinant Human CD43/Sialophorin Protein is derived from E. coli. The Recombinant Human CD43/Sialophorin Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_003114
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol
Gene ID (Entrez) 84687
Molecular Weight (g/mol) 25.8kDa
Name PPP1R9B/Spinophilin Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen SPN, 20-253aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSSTTAVQT PTSGEPLVST SEPLSSKMYT TSITSDPKAD STGDQTSALP PSTSINEGSP LWTSIGASTG SPLPEPTTYQ EVSIKMSSVP QETPHATSHP AVPITANSLG SHTVTGGTIT TNSPETSSRT SGAPVTTAAS SLETSRGTSG PPLTMATVSL ETSKGTSGPP VTMATDSLET STGTTGPPVT MTTGSLEPSS GASGPQVSSV KLSTMMSPTT STNASTVPFR NPDENSR
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.