missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Rheb T7 Protein

Product Code. 18211073 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18211073

Brand: Novus Biologicals™ NBC118435

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal T7-tag and corresponding to the amino acids 1-181 of Human Rheb The Recombinant Human Rheb Protein is derived from E. coli. The Recombinant Human Rheb Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15382
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 1 mM DTT, 10% glycerol
Gene ID (Entrez) 6009
Molecular Weight (g/mol) M.W. (theoretical): 21.7 kDa
Name Rheb Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen RheB, 1-181aa, Human,T7-tagged, Recombinant, E.coli (NP_005605).
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol RHEB
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Cancer, mTOR Pathway, Signal Transduction
Purity or Quality Grade >95%, by SDS-PAGE
Protein Rheb
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.