missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human RPAIN His Protein

Product Code. 18197661 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18197661

Brand: Novus Biologicals™ NBP223447

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-106 of Human RPAIN The Recombinant Human RPAIN Protein is derived from E. coli. The Recombinant Human RPAIN Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001153718
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 40% glycerol, 2mM DTT
Gene ID (Entrez) 84268
Molecular Weight (g/mol) 14.7kDa
Name RPAIN Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen RPAIN, 1-106aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAESLRS PRRSLYKLVG SPPWKEAFRQ RCLERMRNSR DRLLNRYRQA GSSGPGNSQN SFLVQEVMEE EWNALQSVEN CPEDLAQLEE LIDMAVLEEI QQELINQGL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Signal Transduction
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.