missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human SAT1 His Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-171 of Human SAT1 The Recombinant Human SAT1 Protein is derived from E. coli. The Recombinant Human SAT1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_002961 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 10% glycerol |
| Gene ID (Entrez) | 6303 |
| Molecular Weight (g/mol) | 22.1kDa |
| Name | SAT1 Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MAKFVIRPAT AADCSDILRL IKELAKYEYM EEQVILTEKD LLEDGFGEHP FYHCLVAEVP KEHWTPEGHS IVGFAMYYFT YDPWIGKLLY LEDFFVMSDY RGFGIGSEIL KNLSQVAMRC RCSSMHFLVA EWNEPSINFY KRRGASDLSS EEGWRLFKID KEYLLKMATE E |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction