Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 19-122 of Human Serum Amyloid A1 The Recombinant Human Serum Amyloid A1 Protein is derived from E. coli. The Recombinant Human Serum Amyloid A1 Protein has been validated for the following applications: SDS-Page.