missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human TBP like protein TLP His Protein

Product Code. 18150032 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18150032 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18150032 Supplier Novus Biologicals™ Supplier No. NBP222853

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-186 of Human TBP like protein TLP The Recombinant Human TBP like protein TLP Protein is derived from E. coli. The Recombinant Human TBP like protein TLP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_004856
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 50% glycerol, 2mM DTT, 1mM EDTA
Gene ID (Entrez) 9519
Molecular Weight (g/mol) 23.3kDa
Name TBP like protein TLP Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen TBPL1, 1-186aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMDADSDV ALDILITNVV CVFRTRCHLN LRKIALEGAN VIYKRDVGKV LMKLRKPRIT ATIWSSGKII CTGATSEEEA KFGARRLARS LQKLGFQVIF TDFKVVNVLA VCNMPFEIRL PEFTKNNRPH ASYEPELHPA VCYRIKSLRA TLQIFSTGSI TVTGPNVKAV ATAVEQIYPF VFESRKEIL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.