missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human TUSC2 His Protein

Product Code. 18199661 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
This item is not returnable. View return policy

Product Code. 18199661

Brand: Novus Biologicals™ NBP222910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-110 of Human TUSC2 The Recombinant Human TUSC2 Protein is derived from E. coli. The Recombinant Human TUSC2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_009206
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 11334
Molecular Weight (g/mol) 14.5kDa
Name TUSC2 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen TUSC2, 1-110aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMGASGSK ARGLWPFASA AGGGGSEAAG AEQALVRPRG RAVPPFVFTR RGSMFYDEDG DLAHEFYEET IVTKNGQKRA KLRRVHKNLI PQGIVKLDHP RIHVDFPVIL YEV
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.