Learn More
Novus Biologicals™ Recombinant Human VAMP-1 His Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-91 of Human VAMP-1 The Recombinant Human VAMP-1 Protein is derived from E. coli. The Recombinant Human VAMP-1 Protein has been validated for the following applications: SDS-Page.