missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ p38 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-360 of Human p38 The Recombinant Human p38 alpha Protein is derived from E. coli. The Recombinant Human p38 alpha Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 100mM NaCl |
| Molecular Weight (g/mol) | 43.7kDa |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Source | Human |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | p38 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction