missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Raji Whole Cell Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Description
The Raji Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, SDS-Page.
Specifications
Specifications
| For Use With (Application) | SDS-Page, Western Blot |
| Tissue | Blood Lymph |
| Quantity | 0.5 mg |
| Storage Buffer | 1X SDS-PAGE Sample Buffer [0.05mM Tris HCl, 2% SDS, 10% Glycerol, 0.1% bromophenol blue, pH 6.8] |
| Test Specificity | Raji whole cell extract lysate |
| Cellular Fraction | Whole |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction