missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TRPM7 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
A peptide to TRPM7.
Specifications
Specifications
| Gene ID (Entrez) | 54822 |
| Species | Rat |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 500-243. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK |
| Gene Symbol | TRPM7 |
| Product Type | TRPM7 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction