missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOX1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 482.00
Specifications
| Antigen | NOX1 |
|---|---|
| Dilution | Western Blot 1:200 - 1:500, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687040
|
Novus Biologicals
NBP2-93835-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606130
|
Novus Biologicals
NBP2-93835-0.1ml |
0.1 mL |
€ 482.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOX1 Polyclonal antibody specifically detects NOX1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| NOX1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Direct Reversal of DNA Damage, Endocrinology, Immunology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 27035 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200 - 1:500, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 1.6.3, GP91-2, Mitogenic oxidase 1, MOX-1, MOX1NADH/NADPH mitogenic oxidase subunit P65-MOX, NADPH oxidase 1, NADPH oxidase homolog-1, NOH1mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating), NOH-1NADPH oxidase 1 variant NOH-1L, NOX-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 418-564 of human NOX1 (NP_008983.2). WYKFQCADHNLKTKKIYFYWICRETGAFSWFNNLLTSLEQEMEELGKVGFLNYRLFLTGWDSNIVGHAALNFDKATDIVTGLKQKTSFGRPMWDNEFSTIATSHPKSVVGVFLCGPRTLAKSLRKCCHRYSSLDPRKVQFYFNKENF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title