missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114489
This item is not returnable.
View return policy
Description
Positive controls that may be used include: Rat Brain Tissue Lysate.
Mox1 and the glycoprotein gp91-phox are largely related proteins that are essential components of the NADPH oxidase. The superoxide-generating NADPH oxidase complex expresses in phagocytes, neuroepithelial bodies, vascular smooth muscle cells, and endothelial cells. It includes a membrane bound flavocytochrome containing two subunits, gp91-phox and p22-phox, and the cytosolic proteins p47-phox and p67-phox. The p22- and gp91-phox subunits also function as surface O2 sensors that initiate cellular signaling in response to hypoxic conditions. Mox1 and gp91 contain identical C-terminal sequence identity, yet possess distinct expression patterns. gp91-phox expresses in eosinophils, neutrophils, monocytes, and B-lymphocytes, whereas Mox1 is predominantly detected in the colon, with low expression in the uterus and prostate.
Specifications
| NOX2 | |
| Polyclonal | |
| Unconjugated | |
| Cybb | |
| AMCBX2; C88302; Cgd; CGD91-phox; Cybb; Cyd; cytochrome b(558) subunit beta; cytochrome b-245 beta chain; cytochrome b-245 beta polypeptide; cytochrome b-245 heavy chain; cytochrome b-245, beta polypeptide; cytochrome b558 subunit beta; endothelial type gp91-phox; GP91 PHOX; GP911; GP91-1; GP91PHOX; gp91-phox; Heme-binding membrane glycoprotein gp91phox; IMD34; NADPH oxidase 2; neutrophil cytochrome b 91 kDa polypeptide; Nox2; p22 phagocyte B-cytochrome; p91PHOX; p91-PHOX; superoxide-generating NADPH oxidase heavy chain subunit | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13058, 1536, 66021 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P04839, Q61093 | |
| Cybb | |
| Rabbit polyclonal NOX2 antibody was raised against a 15 amino acid peptide near the amino terminus of human NOX2. The immunogen is located within amino acids 140 - 190 of NOX2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction